MilliporeSigma
  • Simultaneous determination of three herbicides by differential pulse voltammetry and chemometrics.
Simultaneous determination of three herbicides by differential pulse voltammetry and chemometrics.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2011-04-23)
Yongnian Ni, Lin Wang, Serge Kokot
ABSTRACT
A novel differential pulse voltammetry method (DPV) was researched and developed for the simultaneous determination of Pendimethalin, Dinoseb and sodium 5-nitroguaiacolate (5NG) with the aid of chemometrics. The voltammograms of these three compounds overlapped significantly, and to facilitate the simultaneous determination of the three analytes, chemometrics methods were applied. These included classical least squares (CLS), principal component regression (PCR), partial least squares (PLS) and radial basis function-artificial neural networks (RBF-ANN). A separately prepared verification data set was used to confirm the calibrations, which were built from the original and first derivative data matrices of the voltammograms. On the basis relative prediction errors and recoveries of the analytes, the RBF-ANN and the DPLS (D - first derivative spectra) models performed best and are particularly recommended for application. The DPLS calibration model was applied satisfactorily for the prediction of the three analytes from market vegetables and lake water samples.
Product Description
Supelco
Dinoseb, PESTANAL®, analytical standard
Supelco
Dinoseb, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Pendimethalin solution, 100 μg/mL in methanol, PESTANAL®, analytical standard
Supelco
Pendimethalin, PESTANAL®, analytical standard