MilliporeSigma
  • Determination of 60 pesticides in hen eggs using the QuEChERS procedure followed by LC-MS/MS and GC-MS/MS.
Determination of 60 pesticides in hen eggs using the QuEChERS procedure followed by LC-MS/MS and GC-MS/MS.
Food chemistry (2019-07-02)
Nho-Eul Song, Jun Young Lee, Ahmad Rois Mansur, Hae Won Jang, Min-Cheol Lim, Yunyeol Lee, Miyoung Yoo, Tae Gyu Nam
ABSTRACT
An analytical method involving QuEChERS (quick, easy, cheap, effective, rugged, and safe) sample preparation, followed by LC-MS/MS and GC-MS/MS was developed and validated for the determination of 60 pesticides in eggs. Recoveries of 70-120% were achieved for selected pesticides and relative standard deviations <20% were obtained for most analytes at three concentrations. The limit of quantification was <10 µg kg-1 for 83% of the total pesticides. This method was used to analyze 58 egg samples and the residues of seven pesticides (disulfoton, fipronil sulfone, cyromazine, o,p-DDT, p,p-DDD, p,p-DDT, and permethrin) were quantified in 16 egg samples at levels of 5-10 µg kg-1, which was below the corresponding the maximum residue levels, as established by Korean Ministry of Food and Drug Safety. We demonstrated that LC-MS/MS and GC-MS/MS in combination with QuEChERS can be used to routinely monitor multiple pesticide residues in egg samples.
Product Description
Supelco
Disulfoton-sulfone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Clofentezine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland