MilliporeSigma
  • Quantification of methyldopa in human plasma by high-performance liquid chromatography-electrospray tandem mass spectrometry application to a bioequivalence study.
Quantification of methyldopa in human plasma by high-performance liquid chromatography-electrospray tandem mass spectrometry application to a bioequivalence study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2002-03-13)
Celso H Oliveira, Rafael E Barrientos-Astigarraga, Mauro Sucupira, Gustavo S Graudenz, Marcelo N Muscará, Nucci Gilberto De
ABSTRACT
A method based on LC-MS-MS is described for the determination of methyldopa in human plasma using dopa-phenyl-D3 as the internal standard. The method has a chromatographic run time of 5.5 min and was linear in the range of 20-5000 ng/ml. The limit of quantitation was 20 ng/ml, the intra-day precisions were 7.3, 5.4 and 4.3% and the intra-day accuracies were -8.0, -1.3 and -2.0% for 30, 600 and 3000 ng/ml, respectively. The inter-day precisions were 7.7, 0.5 and 0.7% and the inter-day accuracies were 0.2, -1.1 and -2.3%, respectively, for the above concentrations. This method was employed in a bioequivalence study of two tablet formulations of methyldopa.
Sigma-Aldrich
L-Dopa-(phenyl-d3), ≥98 atom % D, ≥98% (CP)