MilliporeSigma
  • EGF-mediated suppression of cell extrusion during mucosal damage attenuates opportunistic fungal invasion.
EGF-mediated suppression of cell extrusion during mucosal damage attenuates opportunistic fungal invasion.
Cell reports (2021-03-25)
Sebastian Wurster, Oscar E Ruiz, Krystin M Samms, Alexander M Tatara, Nathaniel D Albert, Philip H Kahan, Anh Trinh Nguyen, Antonios G Mikos, Dimitrios P Kontoyiannis, George T Eisenhoffer
ABSTRACT
Severe and often fatal opportunistic fungal infections arise frequently following mucosal damage caused by trauma or cytotoxic chemotherapy. Interaction of fungal pathogens with epithelial cells that comprise mucosae is a key early event associated with invasion, and, therefore, enhancing epithelial defense mechanisms may mitigate infection. Here, we establish a model of mold and yeast infection mediated by inducible epithelial cell loss in larval zebrafish. Epithelial cell loss by extrusion promotes exposure of laminin associated with increased fungal attachment, invasion, and larval lethality, whereas fungi defective in adherence or filamentation have reduced virulence. Transcriptional profiling identifies significant upregulation of the epidermal growth factor receptor ligand epigen (EPGN) upon mucosal damage. Treatment with recombinant human EPGN suppresses epithelial cell extrusion, leading to reduced fungal invasion and significantly enhanced survival. These data support the concept of augmenting epithelial restorative capacity to attenuate pathogenic invasion of fungi associated with human disease.
Sigma-Aldrich
SKI 5C, ≥98% (HPLC)
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
FIBRONECTIN FROM HUMAN PLASMA, liquid, 0.1% (Solution), BioReagent, suitable for cell culture
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
AG 1478, A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.
Sigma-Aldrich
Human Laminin (pepsinized) Purified Protein