MilliporeSigma
  • Chemoproteomics-enabled discovery of covalent RNF114-based degraders that mimic natural product function.
Chemoproteomics-enabled discovery of covalent RNF114-based degraders that mimic natural product function.
Cell chemical biology (2021-01-30)
Mai Luo, Jessica N Spradlin, Lydia Boike, Bingqi Tong, Scott M Brittain, Jeffrey M McKenna, John A Tallarico, Markus Schirle, Thomas J Maimone, Daniel K Nomura
ABSTRACT
The translation of functionally active natural products into fully synthetic small-molecule mimetics has remained an important process in medicinal chemistry. We recently discovered that the terpene natural product nimbolide can be utilized as a covalent recruiter of the E3 ubiquitin ligase RNF114 for use in targeted protein degradation-a powerful therapeutic modality within modern-day drug discovery. Using activity-based protein profiling-enabled covalent ligand-screening approaches, here we report the discovery of fully synthetic RNF114-based recruiter molecules that can also be exploited for PROTAC applications, and demonstrate their utility in degrading therapeutically relevant targets, such as BRD4 and BCR-ABL, in cells. The identification of simple and easily manipulated drug-like scaffolds that can mimic the function of a complex natural product is beneficial in further expanding the toolbox of E3 ligase recruiters, an area of great importance in drug discovery and chemical biology.
Sigma-Aldrich
Anti-RNF114 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution