MilliporeSigma
  • Effects of 1800 MHz radiofrequency fields on signal transduction and antioxidant proteins in human A172 glioblastoma cells.
Effects of 1800 MHz radiofrequency fields on signal transduction and antioxidant proteins in human A172 glioblastoma cells.
International journal of radiation biology (2021-05-29)
James P McNamee, Veronica S Grybas, Sami S Qutob, Pascale V Bellier
ABSTRACT
To assess the effects of 1800 MHz radiofrequency electromagnetic field (RF-EMF) exposure on the expression of signal transduction and antioxidant proteins in a human-derived A172 glioblastoma cell line. Adherent human-derived A172 glioblastoma cells (1.0 × 105 cells per 35 mm culture dish, containing 2 mL DMEM media) were exposed to 1800 MHz continuous-wave (CW) or GSM-modulated RF fields, in the presence or absence of serum for 5, 30 or 240 min at a specific absorption rate (SAR) of 0 (sham) or 2.0 W/kg. Concurrent negative (vehicle) and positive controls (1 µg/mL anisomycin) were included in each experiment. Cell lysates were collected immediately after exposure, stabilized by protease and phosphatase inhibitors in lysis buffer, then frozen and maintained at -80 °C until analysis. The relative expression levels of phosphorylated- and total-signal transduction proteins (CREB, JNK, NF-κB, ERK1/2, Akt, p70S6K, STAT3 and STAT5) and antioxidant proteins (SOD1, SOD2, CAT, TRX1, PRX2) were assessed using Milliplex magnetic bead array panels and a MagPix Multiplex imaging system. In cells exposed to 1800 MHz continuous-wave RF-EMF with the presence of serum in the culture medium, CAT expression was statistically significantly decreased after a 30 min exposure, total JNK was decreased at both 30 and 240 min of exposure, STAT3 was decreased after 240 min of exposure and phosphorylated-CREB expression was decreased after 30 min of exposure. In cells exposed to 1800 MHz GSM-modulated RF-EMF in serum-free cultures, the expression level of total STAT5 was decreased after 30 and 240 min of exposure. These observed changes were detected sporadically across time-points, culture conditions and RF-EMF exposure conditions indicating the likelihood of false positive events. When cells were treated with anisomycin for 15 min as a positive control, dramatic increases in the expression of phosphorylated signaling proteins were observed in both serum-starved and serum-fed A172 cells, with larger fold change increases in the serum-free cultures. No statistically significant differences in the expression levels of SOD1, SOD2 or TRX1 were observed under any tested conditions after exposure to RF-EMF. The current study found no consistent evidence of changes in the expression of antioxidant proteins (SOD1, SOD2, CAT or TRX2) or a variety of signal transductions proteins (CREB, JNK, NF-κB, ERK1/2, Akt, p70S6K, STAT3, STAT5) in a human-derived glioblastoma A172 cell line in response to exposure to 1800 MHz continuous-wave or GSM-modulated RF-EMF for 5, 30 or 240 min in either serum-free or serum-containing cultures.
Product Description
Millipore
MILLIPLEX MAP Multi-Pathway Magnetic Bead 9-Plex - Cell Signaling Multiplex Assay
Millipore
MILLIPLEX® Multi-Pathway Total Magnetic Bead 9-Plex - Cell Signaling Multiplex Assay