MilliporeSigma
  • A missense mutation in a patient with developmental delay affects the activity and structure of the hexosamine biosynthetic pathway enzyme AGX1.
A missense mutation in a patient with developmental delay affects the activity and structure of the hexosamine biosynthetic pathway enzyme AGX1.
FEBS letters (2020-10-25)
Xiping Chen, Olawale G Raimi, Andrew T Ferenbach, Daan M F van Aalten
ABSTRACT
O-GlcNAcylation is a post-translational modification catalysed by O-GlcNAc transferase (OGT). Missense mutations in OGT have been associated with developmental disorders, OGT-linked congenital disorder of glycosylation (OGT-CDG), which are characterized by intellectual disability. OGT relies on the hexosamine biosynthetic pathway (HBP) for provision of its UDP-GlcNAc donor. We considered whether mutations in UDP-N-acetylhexosamine pyrophosphorylase (UAP1), which catalyses the final step in the HBP, would phenocopy OGT-CDG mutations. A de novo mutation in UAP1 (NM_001324114:c.G685A:p.A229T) was reported in a patient with intellectual disability. We show that this mutation is pathogenic and decreases the stability and activity of the UAP1 isoform AGX1 in vitro. X-ray crystallography reveals a structural shift proximal to the mutation, leading to a conformational change of the N-terminal domain. These data suggest that the UAP1A229T missense mutation could be a contributory factor to the patient phenotype.
Sigma-Aldrich
Uridine 5′-triphosphate trisodium salt hydrate, from yeast, Type III, ≥96%