MilliporeSigma
  • The tumor suppressor MIR139 is silenced by POLR2M to promote AML oncogenesis.
The tumor suppressor MIR139 is silenced by POLR2M to promote AML oncogenesis.
Leukemia (2021-11-07)
Christiaan J Stavast, Iris van Zuijen, Elena Karkoulia, Arman Özçelik, Antoinette van Hoven-Beijen, Leticia G Leon, Jane S A Voerman, George M C Janssen, Peter A van Veelen, Monika Burocziova, Rutger W W Brouwer, Wilfred F J van IJcken, Alex Maas, Eric M Bindels, Vincent H J van der Velden, Christopher Schliehe, Peter D Katsikis, Meritxell Alberich-Jorda, Stefan J Erkeland
ABSTRACT
MIR139 is a tumor suppressor and is commonly silenced in acute myeloid leukemia (AML). However, the tumor-suppressing activities of miR-139 and molecular mechanisms of MIR139-silencing remain largely unknown. Here, we studied the poorly prognostic MLL-AF9 fusion protein-expressing AML. We show that MLL-AF9 expression in hematopoietic precursors caused epigenetic silencing of MIR139, whereas overexpression of MIR139 inhibited in vitro and in vivo AML outgrowth. We identified novel miR-139 targets that mediate the tumor-suppressing activities of miR-139 in MLL-AF9 AML. We revealed that two enhancer regions control MIR139 expression and found that the polycomb repressive complex 2 (PRC2) downstream of MLL-AF9 epigenetically silenced MIR139 in AML. Finally, a genome-wide CRISPR-Cas9 knockout screen revealed RNA Polymerase 2 Subunit M (POLR2M) as a novel MIR139-regulatory factor. Our findings elucidate the molecular control of tumor suppressor MIR139 and reveal a role for POLR2M in the MIR139-silencing mechanism, downstream of MLL-AF9 and PRC2 in AML. In addition, we confirmed these findings in human AML cell lines with different oncogenic aberrations, suggesting that this is a more common oncogenic mechanism in AML. Our results may pave the way for new targeted therapy in AML.
Sigma-Aldrich
SGC0946, ≥98% (HPLC)
Sigma-Aldrich
UNC1999, ≥98% (HPLC)