MilliporeSigma
Pyrrolidine PNA: a novel conformationally restricted PNA analogue.
Organic letters (2001-01-11)
A Püschl, T Tedeschi, P E Nielsen
ABSTRACT
[reaction:see text] A new conformationally restricted PNA adenine monomer has been synthesized in 13 steps from cis-4-hydroxy-D-proline. A fully modified adenine decamer displayed improved binding affinity toward complementary DNA and RNA oligonucleotides as compared to that of the parent PNA adenine decamer.
Sigma-Aldrich
cis-4-Hydroxy-D-proline