MilliporeSigma
  • Large-scale screening identifies a novel microRNA, miR-15a-3p, which induces apoptosis in human cancer cell lines.
Large-scale screening identifies a novel microRNA, miR-15a-3p, which induces apoptosis in human cancer cell lines.
RNA biology (2013-01-29)
Aliaksandr Druz, Yu-Chi Chen, Rajarshi Guha, Michael Betenbaugh, Scott E Martin, Joseph Shiloach
ABSTRACT
MicroRNAs (miRNAs) have been found to be involved in cancer initiation, progression and metastasis and, as such, have been suggested as tools for cancer detection and therapy. In this work, a large-scale screening of the complete miRNA mimics library demonstrated that hsa-miR-15a-3p had a pro-apoptotic role in the following human cancer cells: HeLa, AsPc-1, MDA-MB-231, KB3, ME180, HCT-116 and A549. MiR-15a-3p is a novel member of the pro-apoptotic miRNA cluster, miR-15a/16, which was found to activate Caspase-3/7 and to cause viability loss in B/CMBA.Ov cells during preliminary screening. Subsequent microarrays and bioinformatics analyses identified the following four anti-apoptotic genes: bcl2l1, naip5, fgfr2 and mybl2 as possible targets for the mmu-miR-15a-3p in B/CMBA.Ov cells. Follow-up studies confirmed the pro-apoptotic role of hsa-miR-15a-3p in human cells by its ability to activate Caspase-3/7, to reduce cell viability and to inhibit the expression of bcl2l1 (bcl-xL) in HeLa and AsPc-1 cells. MiR-15-3p was also found to reduce viability in HEK293, MDA-MB-231, KB3, ME180, HCT-116 and A549 cell lines and, therefore, may be considered for apoptosis modulating therapies in cancers associated with high Bcl-xL expression (cervical, pancreatic, breast, lung and colorectal carcinomas). The capability of hsa-miR-15a-3p to induce apoptosis in these carcinomas may be dependent on the levels of Bcl-xL expression. The use of endogenous inhibitors of bcl-xL and other anti-apoptotic genes such as hsa-miR-15a-3p may provide improved options for apoptosis-modulating therapies in cancer treatment compared with the use of artificial antisense oligonucleotides.
Sigma-Aldrich
4-Ethylmorpholine, ≥97.0% (GC)