MilliporeSigma
  • Noninvasive neuroelectronic interfacing with synaptically connected snail neurons immobilized on a semiconductor chip.
Noninvasive neuroelectronic interfacing with synaptically connected snail neurons immobilized on a semiconductor chip.
Proceedings of the National Academy of Sciences of the United States of America (2001-08-30)
G Zeck, P Fromherz
ABSTRACT
A hybrid circuit of a semiconductor chip and synaptically connected neurons was implemented and characterized. Individual nerve cells from the snail Lymnaea stagnalis were immobilized on a silicon chip by microscopic picket fences of polyimide. The cells formed a network with electrical synapses after outgrowth in brain conditioned medium. Pairs of neurons were electronically interfaced for noninvasive stimulation and recording. Voltage pulses were applied to a capacitive stimulator on the chip to excite the attached neuron. Signals were transmitted in the neuronal net and elicited an action potential in a second neuron. The postsynaptic excitation modulated the current of a transistor on the chip. The implementation of the silicon-neuron-neuron-silicon circuit constitutes a proof-of-principle experiment for the development of neuroelectronic systems to be used in studies on neuronal signal processing, neurocomputation, and neuroprosthetics.
Sigma-Aldrich
Gentamicin sulfate salt, potency: ≥590 I.U. Gentamicin base per mg