MilliporeSigma
  • SSSPTA is essential for serine palmitoyltransferase function during development and hematopoiesis.
SSSPTA is essential for serine palmitoyltransferase function during development and hematopoiesis.
The Journal of biological chemistry (2021-03-05)
Velayoudame Parthibane, Jing Lin, Diwash Acharya, Thiruvaimozhi Abimannan, Sargur Madabushi Srideshikan, Kimberly Klarmann, Acong Yang, Ferri Soheilian, Kunio Nagashima, Stephen D Fox, Thorkell Andresson, Lino Tessarollo, Jonathan R Keller, Usha Acharya, Jairaj K Acharya
ABSTRACT
Serine palmitoyltransferase complex (SPT) mediates the first and rate-limiting step in the de novo sphingolipid biosynthetic pathway. The larger subunits SPTLC1 and SPTLC2/SPTLC3 together form the catalytic core while a smaller third subunit either SSSPTA or SSSPTB has been shown to increase the catalytic efficiency and provide substrate specificity for the fatty acyl-CoA substrates. The in vivo biological significance of these smaller subunits in mammals is still unknown. Here, using two null mutants, a conditional null for ssSPTa and a null mutant for ssSPTb, we show that SSSPTA is essential for embryogenesis and mediates much of the known functions of the SPT complex in mammalian hematopoiesis. The ssSPTa null mutants are embryonic lethal at E6.5 much like the Sptlc1 and Sptlc2 null alleles. Mx1-Cre induced deletion of ssSPTa leads to lethality and myelopoietic defect. Chimeric and competitive bone marrow transplantation experiments show that the defect in myelopoiesis is accompanied by an expansion of the Lin-Sca1+c-Kit+ stem and progenitor compartment. Progenitor cells that fail to differentiate along the myeloid lineage display evidence of endoplasmic reticulum stress. On the other hand, ssSPTb null mice are homozygous viable, and analyses of the bone marrow cells show no significant difference in the proliferation and differentiation of the adult hematopoietic compartment. SPTLC1 is an obligatory subunit for the SPT function, and because Sptlc1-/- and ssSPTa-/- mice display similar defects during development and hematopoiesis, we conclude that an SPT complex that includes SSSPTA mediates much of its developmental and hematopoietic functions in a mammalian model.
Sigma-Aldrich
Palmitoyl coenzyme A lithium salt, ≥90%