MilliporeSigma
  • Drug Resistance Mutations Among South African Children Living With HIV on WHO-recommended ART Regimens.
Drug Resistance Mutations Among South African Children Living With HIV on WHO-recommended ART Regimens.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2020-08-01)
Stephanie Hackett, Chloe A Teasdale, Sherri Pals, Anthony Muttiti, Mary Mogashoa, Joy Chang, Clement Zeh, Artur Ramos, Emilia D Rivadeneira, Joshua DeVos, Katrina Sleeman, Elaine J Abrams
ABSTRACT
Children living with human immunodeficiency virus (HIV) (CLHIV) receiving antiretroviral therapy (ART) in resource-limited settings are susceptible to high rates of acquired HIV drug resistance (HIVDR), but few studies include children initiating age-appropriate World Health Organization (WHO)-recommended first-line regimens. We report data from a cohort of ART-naive South African children who initiated first-line ART. ART-eligible CLHIV aged 0-12 years were enrolled from 2012 to 2014 at 5 public South African facilities and were followed for up to 24 months. Enrolled CLHIV received standard-of-care WHO-recommended first-line ART. At the final study visit, a dried blood spot sample was obtained for viral load and genotypic resistance testing. Among 72 successfully genotyped CLHIV, 49 (68.1%) received ABC/3TC/LPV/r, and 23 (31.9%) received ABC/3TC/EFV. All but 2 children on ABC/3TC/LPV/r were <3 years, and all CLHIV on ABC/3TC/EFV were ≥3 years. Overall, 80.6% (58/72) had at least one drug resistance mutation (DRM). DRMs to nonnucleoside reverse transcriptase inhibitors (NNRTIs) and nucleoside reverse transcriptase inhibitors (NRTIs) were found among 65% and 51% of all CLHIV, respectively, with no statistical difference by ART regimen. More CLHIV on ABC/3TC/EFV, 47.8% (11/23), were found to have 0 or only 1 effective antiretroviral drug remaining in their current regimen compared to 8.2% (4/49) on ABC/3TC/LPV/r. High levels of NNRTI and NRTI DRMs among CLHIV receiving ABC/3TC/LPV/r suggests a lasting impact of failed mother-to-child transmission interventions on DRMs. However, drug susceptibility analysis reveals that CLHIV with detectable viremia on ABC/3TC/LPV/r are more likely to have maintained at least 2 effective agents on their current HIV regimen than those on ABC/3TC/EFV.
Sigma-Aldrich
7-Amino-4-hydroxy-2-naphthalenesulfonic acid, technical, ≥90% (T)
Supelco
4-Aminoantipyrine, for spectrophotometric det. of H2O2 and phenols, ≥98.0%