MilliporeSigma
  • BRAFV600E induces reversible mitotic arrest in human melanocytes via microrna-mediated suppression of AURKB.
BRAFV600E induces reversible mitotic arrest in human melanocytes via microrna-mediated suppression of AURKB.
eLife (2021-11-24)
Andrew S McNeal, Rachel L Belote, Hanlin Zeng, Marcus Urquijo, Kendra Barker, Rodrigo Torres, Meghan Curtin, A Hunter Shain, Robert Hi Andtbacka, Sheri Holmen, David H Lum, Timothy H McCalmont, Matt W VanBrocklin, Douglas Grossman, Maria L Wei, Ursula E Lang, Robert L Judson-Torres
ABSTRACT
Benign melanocytic nevi frequently emerge when an acquired BRAFV600E mutation triggers unchecked proliferation and subsequent arrest in melanocytes. Recent observations have challenged the role of oncogene-induced senescence in melanocytic nevus formation, necessitating investigations into alternative mechanisms for the establishment and maintenance of proliferation arrest in nevi. We compared the transcriptomes of melanocytes from healthy human skin, nevi, and melanomas arising from nevi and identified a set of microRNAs as highly expressed nevus-enriched transcripts. Two of these microRNAs-MIR211-5p and MIR328-3p-induced mitotic failure, genome duplication, and proliferation arrest in human melanocytes through convergent targeting of AURKB. We demonstrate that BRAFV600E induces a similar proliferation arrest in primary human melanocytes that is both reversible and conditional. Specifically, BRAFV600E expression stimulates either arrest or proliferation depending on the differentiation state of the melanocyte. We report genome duplication in human melanocytic nevi, reciprocal expression of AURKB and microRNAs in nevi and melanomas, and rescue of arrested human nevus cells with AURKB expression. Taken together, our data describe an alternative molecular mechanism for melanocytic nevus formation that is congruent with both experimental and clinical observations.
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
MCDB 153 Medium, With trace elements, L-glutamine and 28mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture