MilliporeSigma
  • 5'-Modifications improve potency and efficacy of DNA donors for precision genome editing.
5'-Modifications improve potency and efficacy of DNA donors for precision genome editing.
eLife (2021-10-20)
Krishna S Ghanta, Zexiang Chen, Aamir Mir, Gregoriy A Dokshin, Pranathi M Krishnamurthy, Yeonsoo Yoon, Judith Gallant, Ping Xu, Xiao-Ou Zhang, Ahmet Rasit Ozturk, Masahiro Shin, Feston Idrizi, Pengpeng Liu, Hassan Gneid, Alireza Edraki, Nathan D Lawson, Jaime A Rivera-Pérez, Erik J Sontheimer, Jonathan K Watts, Craig C Mello
ABSTRACT
Nuclease-directed genome editing is a powerful tool for investigating physiology and has great promise as a therapeutic approach to correct mutations that cause disease. In its most precise form, genome editing can use cellular homology-directed repair (HDR) pathways to insert information from an exogenously supplied DNA-repair template (donor) directly into a targeted genomic location. Unfortunately, particularly for long insertions, toxicity and delivery considerations associated with repair template DNA can limit HDR efficacy. Here, we explore chemical modifications to both double-stranded and single-stranded DNA-repair templates. We describe 5'-terminal modifications, including in its simplest form the incorporation of triethylene glycol (TEG) moieties, that consistently increase the frequency of precision editing in the germlines of three animal models (Caenorhabditis elegans, zebrafish, mice) and in cultured human cells.
Product Description
Roche
CDP-Star®, ready-to-use, >98%, solution, suitable for dot blot, suitable for Northern blotting, suitable for Southern blotting
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
DIG Wash and Block Buffer Set, storage temp.:2-8°C
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Roche
PCR DIG Labeling Mix, solution, suitable for PCR