MilliporeSigma
  • The Lyme disease agent co-opts adiponectin receptor-mediated signaling in its arthropod vector.
The Lyme disease agent co-opts adiponectin receptor-mediated signaling in its arthropod vector.
eLife (2021-11-17)
Xiaotian Tang, Yongguo Cao, Gunjan Arora, Jesse Hwang, Andaleeb Sajid, Courtney L Brown, Sameet Mehta, Alejandro Marín-López, Yu-Min Chuang, Ming-Jie Wu, Hongwei Ma, Utpal Pal, Sukanya Narasimhan, Erol Fikrig
ABSTRACT
Adiponectin-mediated pathways contribute to mammalian homeostasis; however, little is known about adiponectin and adiponectin receptor signaling in arthropods. In this study, we demonstrate that Ixodes scapularis ticks have an adiponectin receptor-like protein (ISARL) but lack adiponectin, suggesting activation by alternative pathways. ISARL expression is significantly upregulated in the tick gut after Borrelia burgdorferi infection, suggesting that ISARL signaling may be co-opted by the Lyme disease agent. Consistent with this, RNA interference (RNAi)-mediated silencing of ISARL significantly reduced the B. burgdorferi burden in the tick. RNA-seq-based transcriptomics and RNAi assays demonstrate that ISARL-mediated phospholipid metabolism by phosphatidylserine synthase I is associated with B. burgdorferi survival. Furthermore, the tick complement C1q-like protein 3 interacts with ISARL, and B. burgdorferi facilitates this process. This study identifies a new tick metabolic pathway that is connected to the life cycle of the Lyme disease spirochete.
Sigma-Aldrich
BSK-H Medium, Complete, sterile-filtered, with 6% rabbit serum, suitable for Borrelia burgdorferi
Sigma-Aldrich
ADIPONECTIN mouse, recombinant, expressed in Hi-5 Insect cells, ≥95% (SDS-PAGE), ≥95% (HPLC)