MilliporeSigma
Soil urease inhibition by various plant extracts.
PloS one (2021-10-15)
Muhammad Ajmal Rana, Rashid Mahmood, Sajid Ali
ABSTRACT
Urea is the most popular and widely used nitrogenous fertilizer. High soil urease activity rapidly hydrolyses applied urea to ammonia which contributes to soil nitrogen (N) losses and reduces N use efficiency of crop plants. The ammonia losses can be minimized by the inhibition of soil urease activity which has been explored using various potential chemical inhibitors. However, the soil urease activity inhibition potential of plant extracts is rarely explored to date. In the present study, extracts of 35 plant materials were taken and evaluated against jack bean urease. Eleven extracts, showing >50% jack bean urease inhibition, were selected and further investigated in 13 soils collected from various districts of Punjab, Pakistan. Interestingly, except Capsicum annum, Melia azedarach, Citrus reticulata and Quercus infectoria, the plant extracts showed urease inhibition activities in soils, the extent of which was lower as compared to that observed in jack bean urease though. Maximum urea hydrolysis inhibition (70%) was noted with Vachellia nilotica which was 40% more than that of hydroquinone (50%) followed by that of Eucalyptus camaldulensis (24%). The extracts of V. nilotica and E. camaldulensis were coated on urea and applied to soil in the next step. At 21st day, 239% and 116% more urea-N was recovered from soil treated with V. nilotica and E. camaldulensis extracts coated urea, respectively, as compared to uncoated urea. Conclusively, these results indicated that the coating of V. nilotica and E. camaldulensis extracts on urea prills prolonged urea persistence in soil owing to minimum urea hydrolysis, probably, the extracts of V. nilotica and E. camaldulensis showed their urease inhibition potential. The results of this study provide a base line for the identification of new soil urease inhibitor compounds from plant materials in future.
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean)