MilliporeSigma
  • The HSP90/R2TP assembly chaperone promotes cell proliferation in the intestinal epithelium.
The HSP90/R2TP assembly chaperone promotes cell proliferation in the intestinal epithelium.
Nature communications (2021-08-12)
Chloé Maurizy, Claire Abeza, Bénédicte Lemmers, Monica Gabola, Ciro Longobardi, Valérie Pinet, Marina Ferrand, Conception Paul, Julie Bremond, Francina Langa, François Gerbe, Philippe Jay, Céline Verheggen, Nicola Tinari, Dominique Helmlinger, Rossano Lattanzio, Edouard Bertrand, Michael Hahne, Bérengère Pradet-Balade
ABSTRACT
The R2TP chaperone cooperates with HSP90 to integrate newly synthesized proteins into multi-subunit complexes, yet its role in tissue homeostasis is unknown. Here, we generated conditional, inducible knock-out mice for Rpap3 to inactivate this core component of R2TP in the intestinal epithelium. In adult mice, Rpap3 invalidation caused destruction of the small intestinal epithelium and death within 10 days. Levels of R2TP substrates decreased, with strong effects on mTOR, ATM and ATR. Proliferative stem cells and progenitors deficient for Rpap3 failed to import RNA polymerase II into the nucleus and they induced p53, cell cycle arrest and apoptosis. Post-mitotic, differentiated cells did not display these alterations, suggesting that R2TP clients are preferentially built in actively proliferating cells. In addition, high RPAP3 levels in colorectal tumors from patients correlate with bad prognosis. Here, we show that, in the intestine, the R2TP chaperone plays essential roles in normal and tumoral proliferation.
Sigma-Aldrich
Anti-NOP58 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-RPAP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer