MilliporeSigma
  • Utility of Immunohistochemistry and Western Blot in Profiling Clinically Suspected Cases of Congenital Muscular Dystrophy.
Utility of Immunohistochemistry and Western Blot in Profiling Clinically Suspected Cases of Congenital Muscular Dystrophy.
Annals of Indian Academy of Neurology (2021-07-06)
Radhika Mhatre, Deepha Sekar, Jessiena Ponmalar, Madhu Nagappa, Preethish-Kumar Veeramani, Kiran Polavarapu, Seena Vengalil, Nalini Atchayaram, Gayathri Narayanappa
ABSTRACT
Immunocharacterization of congenital muscular dystrophy (CMD) to determine the frequency of various subtypes in a large Indian Cohort. This retrospective (2014-2017) study was carried on muscle biopsies of clinically suspected cases of CMD with histological evidence of dystrophy/myopathic features. Immunohistochemistry (IHC) to antibodies against laminin (α2, α5,β1,γ1), Collagen-VI (A1,2,3), and Western blot (WB) for α-dystroglycan and POMT1 was performed. The study included 57 cases, of which 15 cases (26.3%) had mean age at presentation of 3.5 years, M: F = 1.5:1, elevated creatinine kinase (CK) (mean 1657 U/L), global developmental delay, multiple contractures, abnormal facies, white matter hyperintensities and showed laminin-α2 deficiency (Merosin deficient CMD). In addition, secondary reduction in laminin-β1, over-expression of laminin-α5, and preserved laminin-γ1 was noted. Ullrich CMD constituted 11/57 cases (19.2%) with mean age at presentation of 5.3 years, M: F = 1.2:1 and normal CK. They presented with proximal muscle weakness, soft velvety palms and soles, contractures, and joint hyperextensibility. Collagen-VI (A1,2,3) showed either complete (n = 3) or sarcolemmal specific (n = 8) loss of staining. Out of the remaining 31 cases, WB for α-dystroglycan was performed in 17 cases which showed deficiency in seven (12.3%). Three of these in addition revealed secondary partial loss of laminin-α2. WB for POMT1 showed deficiency in a single case clinically diagnosed Walker-Warburg syndrome, who presented with seizures and classical features of pachygyria, lissencephaly, and cerebellar cyst on MRI. Twenty-four cases (42.2%) remained uncharacterized and need genetic evaluation. The study helped in characterizing 57.8% of the proband. Immunotyping helps to direct mutational analysis for targeted genes and offers a potential route for prenatal diagnosis.
Sigma-Aldrich
Anti-Collagen Type VI Antibody, clone 3C4, ascites fluid, clone 3C4, Chemicon®
Sigma-Aldrich
Anti-Laminin γ1 Antibody, clone 2E8, ascites fluid, clone 2E8, Chemicon®
Sigma-Aldrich
Anti-Laminin α5 Antibody, clone 4C7, ascites fluid, clone 4C7, Chemicon®
Sigma-Aldrich
Anti-Collagen Type VI Antibody, clone VI-26, clone VI-26, Chemicon®, from mouse
Sigma-Aldrich
Anti-Laminin α2 Antibody, clone 5H2, ascites fluid, clone 5H2, Chemicon®
Sigma-Aldrich
Anti-Laminin β1 Antibody, clone 4E10, clone 4E10, Chemicon®, from mouse