MilliporeSigma
  • Signatures of plasticity, metastasis, and immunosuppression in an atlas of human small cell lung cancer.
Signatures of plasticity, metastasis, and immunosuppression in an atlas of human small cell lung cancer.
Cancer cell (2021-10-16)
Joseph M Chan, Álvaro Quintanal-Villalonga, Vianne Ran Gao, Yubin Xie, Viola Allaj, Ojasvi Chaudhary, Ignas Masilionis, Jacklynn Egger, Andrew Chow, Thomas Walle, Marissa Mattar, Dig V K Yarlagadda, James L Wang, Fathema Uddin, Michael Offin, Metamia Ciampricotti, Besnik Qeriqi, Amber Bahr, Elisa de Stanchina, Umesh K Bhanot, W Victoria Lai, Matthew J Bott, David R Jones, Arvin Ruiz, Marina K Baine, Yanyun Li, Natasha Rekhtman, John T Poirier, Tal Nawy, Triparna Sen, Linas Mazutis, Travis J Hollmann, Dana Pe'er, Charles M Rudin
ABSTRACT
Small cell lung cancer (SCLC) is an aggressive malignancy that includes subtypes defined by differential expression of ASCL1, NEUROD1, and POU2F3 (SCLC-A, -N, and -P, respectively). To define the heterogeneity of tumors and their associated microenvironments across subtypes, we sequenced 155,098 transcriptomes from 21 human biospecimens, including 54,523 SCLC transcriptomes. We observe greater tumor diversity in SCLC than lung adenocarcinoma, driven by canonical, intermediate, and admixed subtypes. We discover a PLCG2-high SCLC phenotype with stem-like, pro-metastatic features that recurs across subtypes and predicts worse overall survival. SCLC exhibits greater immune sequestration and less immune infiltration than lung adenocarcinoma, and SCLC-N shows less immune infiltrate and greater T cell dysfunction than SCLC-A. We identify a profibrotic, immunosuppressive monocyte/macrophage population in SCLC tumors that is particularly associated with the recurrent, PLCG2-high subpopulation.
Sigma-Aldrich
Anti-RRBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Firefly/Renilla Dual Luciferase Assay, Flash-type dual luciferase assay that allows measurement of both Firefly and Renilla luciferase activity in the same sample with high sensitivity and linearity.
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Anti-PLCG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Xylenes, histological grade