MilliporeSigma
  • SARS-CoV-2 mRNA vaccines induce persistent human germinal centre responses.
SARS-CoV-2 mRNA vaccines induce persistent human germinal centre responses.
Nature (2021-06-29)
Jackson S Turner, Jane A O'Halloran, Elizaveta Kalaidina, Wooseob Kim, Aaron J Schmitz, Julian Q Zhou, Tingting Lei, Mahima Thapa, Rita E Chen, James Brett Case, Fatima Amanat, Adriana M Rauseo, Alem Haile, Xuping Xie, Michael K Klebert, Teresa Suessen, William D Middleton, Pei-Yong Shi, Florian Krammer, Sharlene A Teefey, Michael S Diamond, Rachel M Presti, Ali H Ellebedy
ABSTRACT
SARS-CoV-2 mRNA-based vaccines are about 95% effective in preventing COVID-191-5. The dynamics of antibody-secreting plasmablasts and germinal centre B cells induced by these vaccines in humans remain unclear. Here we examined antigen-specific B cell responses in peripheral blood (n = 41) and draining lymph nodes in 14 individuals who had received 2 doses of BNT162b2, an mRNA-based vaccine that encodes the full-length SARS-CoV-2 spike (S) gene1. Circulating IgG- and IgA-secreting plasmablasts that target the S protein peaked one week after the second immunization and then declined, becoming undetectable three weeks later. These plasmablast responses preceded maximal levels of serum anti-S binding and neutralizing antibodies to an early circulating SARS-CoV-2 strain as well as emerging variants, especially in individuals who had previously been infected with SARS-CoV-2 (who produced the most robust serological responses). By examining fine needle aspirates of draining axillary lymph nodes, we identified germinal centre B cells that bound S protein in all participants who were sampled after primary immunization. High frequencies of S-binding germinal centre B cells and plasmablasts were sustained in these draining lymph nodes for at least 12 weeks after the booster immunization. S-binding monoclonal antibodies derived from germinal centre B cells predominantly targeted the receptor-binding domain of the S protein, and fewer clones bound to the N-terminal domain or to epitopes shared with the S proteins of the human betacoronaviruses OC43 and HKU1. These latter cross-reactive B cell clones had higher levels of somatic hypermutation as compared to those that recognized only the SARS-CoV-2 S protein, which suggests a memory B cell origin. Our studies demonstrate that SARS-CoV-2 mRNA-based vaccination of humans induces a persistent germinal centre B cell response, which enables the generation of robust humoral immunity.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat
Sigma-Aldrich
Mouse Anti-Human IgA Antibody, clone M24A, heavy chain, FITC conjugated, clone M24A, Chemicon®, from mouse