MilliporeSigma
  • Extraction and sequencing of single nuclei from murine skeletal muscles.
Extraction and sequencing of single nuclei from murine skeletal muscles.
STAR protocols (2021-08-13)
Matthieu Dos Santos, Stamatia Gioftsidi, Stéphanie Backer, Léo Machado, Frederic Relaix, Pascal Maire, Philippos Mourikis
ABSTRACT
Single-nucleus RNA sequencing allows the profiling of gene expression in isolated nuclei. Here, we describe a step-by-step protocol optimized for adult mouse skeletal muscles. This protocol provides two main advantages compared to the widely used single-cell protocol. First, it allows us to sequence the myonuclei of the multinucleated myofibers. Second, it circumvents the cell-dissociation-induced transcriptional modifications. For complete details on the use and execution of this protocol, please refer to Dos Santos et al. (2020) and Machado, Geara et al. (2021).
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Trizma® hydrochloride solution, pH 7.4, BioPerformance Certified, 1 M, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin solution, 10% in DPBS, low endotoxin, fatty acid free, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M