MilliporeSigma
  • Stabilization of Collagen Fibrils by Gelatin Addition: A Study of Collagen/Gelatin Dense Phases.
Stabilization of Collagen Fibrils by Gelatin Addition: A Study of Collagen/Gelatin Dense Phases.
Langmuir : the ACS journal of surfaces and colloids (2017-11-01)
François Portier, Claire Teulon, Agnieszka Nowacka-Perrin, Flavien Guenneau, Marie-Claire Schanne-Klein, Gervaise Mosser
ABSTRACT
Collagen and its denatured form, gelatin, are biopolymers of fundamental interest in numerous fields ranging from living tissues to biomaterials, food, and cosmetics. This study aims at characterizing mixtures of those biopolymers at high concentrations (up to 100 mg·mL-1) at which collagen has mesogenic properties. We use a structural approach combining polarization-resolved multiphoton microscopy, polarized light microscopy, magnetic resonance imaging, and transmission electron microscopy to analyze gelatin and collagen/gelatin dense phases in their sol and gel states from the macroscopic to the microscopic scale. We first report the formation of a lyotropic crystal phase of gelatin A and show that gelatin must structure itself in particles to become mesogenic. We demonstrate that mixtures of collagen and gelatin phase segregate, preserving the setting of the pure collagen mesophase at a gelatin ratio of up to 20% and generating a biphasic fractal sample at all tested ratios. Moreover, differential scanning calorimetric analysis shows that each protein separates into two populations. Both populations of gelatins are stabilized by the presence of collagen, whereas only one population of collagen molecules is stabilized by the presence of gelatin, most probably those at the interface of the fibrillated microdomains and of the gelatin phase. Although further studies are needed to fully understand the involved mechanism, these new data should have a direct impact on the bioengineering of those two biopolymers.
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A