MilliporeSigma
  • Differential cellular responses to adhesive interactions with galectin-8- and fibronectin-coated substrates.
Differential cellular responses to adhesive interactions with galectin-8- and fibronectin-coated substrates.
Journal of cell science (2021-03-17)
Wenhong Li, Ana Sancho, Wen-Lu Chung, Yaron Vinik, Jürgen Groll, Yehiel Zick, Ohad Medalia, Alexander D Bershadsky, Benjamin Geiger
ABSTRACT
The mechanisms underlying the cellular response to extracellular matrices (ECMs) that consist of multiple adhesive ligands are still poorly understood. Here, we address this topic by monitoring specific cellular responses to two different extracellular adhesion molecules - the main integrin ligand fibronectin and galectin-8, a lectin that binds β-galactoside residues  - as well as to mixtures of the two proteins. Compared with cell spreading on fibronectin, cell spreading on galectin-8-coated substrates resulted in increased projected cell area, more-pronounced extension of filopodia and, yet, the inability to form focal adhesions and stress fibers. These differences can be partially reversed by experimental manipulations of small G-proteins of the Rho family and their downstream targets, such as formins, the Arp2/3 complex and Rho kinase. We also show that the physical adhesion of cells to galectin-8 was stronger than adhesion to fibronectin. Notably, galectin-8 and fibronectin differently regulate cell spreading and focal adhesion formation, yet act synergistically to upregulate the number and length of filopodia. The physiological significance of the coherent cellular response to a molecularly complex matrix is discussed. This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Myosin IIA, non muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Gly-Arg-Gly-Asp-Ser-Pro-Lys, ≥97% (HPLC)
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder