MilliporeSigma
  • Discovery of novel resorcinol diphenyl ether-based PROTAC-like molecules as dual inhibitors and degraders of PD-L1.
Discovery of novel resorcinol diphenyl ether-based PROTAC-like molecules as dual inhibitors and degraders of PD-L1.
European journal of medicinal chemistry (2020-05-11)
Binbin Cheng, Yichang Ren, Hao Cao, Jianjun Chen
ABSTRACT
Novel resorcinol diphenyl ether-based PROTACs (PROteolysis TArgeting Chimeras) were designed and evaluated for their inhibitory activity against the programmed cell death-1/programmed cell death-ligand 1 (PD-1/PD-L1) pathway and their ability to degrade PD-L1 protein. Most of the compounds displayed excellent inhibitory activities against PD-1/PD-L1, as assessed by the homogenous time-resolved fluorescence (HTRF) binding assay, with IC50 values ranging from 25 nM to 200 nM. Among them, compound P22 is one of the best with an IC50 value of 39.2 nM. In addition to inhibiting PD-1/PD-L1 interaction, P22 also significantly restored the immunity repressed in a co-culture model of Hep3B/OS-8/hPD-L1 and CD3 T cells. Furthermore, flow cytometry (FCM) and western-blot data demonstrated that P22 could moderately reduce the protein levels of PD-L1 in a lysosome-dependent manner, which may contribute to its immune effects. Preliminary FCM and western-blot data suggest that it is possible to build PD-L1-targeting PROTAC-like molecules based on PD-1/PD-L1 small molecule inhibitors, though these compounds showed only modest degradation efficiencies. Collectively, this work suggests that P22 may serve as a starting point for exploring the degradation of PD-L1 by PROTAC-like strategy.
Sigma-Aldrich
5-Amino-Thalidomide, ≥95%