MilliporeSigma
  • A Natural Flavone Tricin from Grains Can Alleviate Tumor Growth and Lung Metastasis in Colorectal Tumor Mice.
A Natural Flavone Tricin from Grains Can Alleviate Tumor Growth and Lung Metastasis in Colorectal Tumor Mice.
Molecules (Basel, Switzerland) (2020-08-23)
Grace Gar-Lee Yue, Si Gao, Julia Kin-Ming Lee, Yuk-Yu Chan, Eric Chun-Wai Wong, Tao Zheng, Xiao-Xiao Li, Pang-Chui Shaw, Monique S J Simmonds, Clara Bik-San Lau
ABSTRACT
Tricin, a flavone isolated from rice bran, has been shown to be chemopreventive in a colorectal cancer (CRC) mouse model. This study aimed to illustrate the inhibitory activities of tricin in colon cancer cells and in a metastatic CRC mouse model. BALB/c mice injected with mouse Colon26-Luc cells into the rectum wall were treated with tricin (37.5 mg/kg) daily for 18 days. Orthotopic colon tumor growth and metastasis to lungs were assessed by in vivo bioluminescence imaging. Results showed that tricin suppressed Colon-Luc cells motility and downregulated phosphorylated Akt, Erk1/2 and NF-κB expressions of human colon cancer HT-29 cells. While tricin treatment suppressed tumor growth and lung metastasis as well as altered the populations of myeloid-derived suppressor cells and regulatory T cells in spleens. In summary, the tumor microenvironment modulatory and anti-metastatic effects of tricin in colon cancer mouse model were shown for the first time, suggesting the potential development of tricin-containing food supplements for CRC patients.
Product Description
Tricin, phyproof® Reference Substance
Sigma-Aldrich
Flavone, ≥99.0%