MilliporeSigma
  • Cascade synthesis and optoelectronic applications of intermediate bandgap Cu3VSe4 nanosheets.
Cascade synthesis and optoelectronic applications of intermediate bandgap Cu3VSe4 nanosheets.
Scientific reports (2020-12-12)
Mimi Liu, Cheng-Yu Lai, Meng Zhang, Daniela R Radu
ABSTRACT
Two-dimensional (2D) ternary materials recently generated interest in optoelectronics and energy-related applications, alongside their binary counterparts. To date, only a few naturally occurring layered 2D ternary materials have been explored. The plethora of benefits owed to reduced dimensionality prompted exploration of expanding non-layered ternary chalcogenides into the 2D realm. This work presents a templating method that uses 2D transition metal dichalcogenides as initiators to be converted into the corresponding ternary chalcogenide upon addition of copper, via a solution-phase synthesis, conducted in high boiling point solvents. The process starts with preparation of VSe2 nanosheets, which are next converted into Cu3VSe4 sulvanite nanosheets (NSs) which retain the 2D geometry while presenting an X-ray diffraction pattern identical with the one for the bulk Cu3VSe4. Both the scanning electron microscopy and transmission microscopy electron microscopy show the presence of quasi-2D morphology. Recent studies of the sulfur-containing sulvanite Cu3VS4 highlight the presence of an intermediate bandgap, associated with enhanced photovoltaic (PV) performance. The Cu3VSe4 nanosheets reported herein exhibit multiple UV-Vis absorption peaks, related to the intermediate bandgaps similar to Cu3VS4 and Cu3VSe4 nanocrystals. To test the potential of Cu3VSe4 NSs as an absorber for solar photovoltaic devices, Cu3VSe4 NSs thin-films deposited on FTO were subjected to photoelectrochemical testing, showing p-type behavior and stable photocurrents of up to ~ 0.036 mA/cm2. The photocurrent shows a ninefold increase in comparison to reported performance of Cu3VSe4 nanocrystals. This proves that quasi-2D sulvanite nanosheets are amenable to thin-film deposition and could show superior PV performance in comparison to nanocrystal thin-films. The obtained electrical impedance spectroscopy signal of the Cu3VSe4 NSs-FTO based electrochemical cell fits an equivalent circuit with the circuit elements of solution resistance (Rs), charge-transfer resistance (Rct), double-layer capacitance (Cdl), and Warburg impedance (W). The estimated charge transfer resistance value of 300 Ω cm2 obtained from the Nyquist plot provides an insight into the rate of charge transfer on the electrode/electrolyte interface.
Sigma-Aldrich
Vanadyl acetylacetonate, 98%