MilliporeSigma
  • Biological evaluation of phenolic constituents from the trunk of Berberis koreana.
Biological evaluation of phenolic constituents from the trunk of Berberis koreana.
Bioorganic & medicinal chemistry letters (2011-03-23)
Ki Hyun Kim, Eunjung Moon, Sang Un Choi, Sun Yeou Kim, Kang Ro Lee
ABSTRACT
A bioassay-guided fractionation and chemical investigation of the trunk of Berberis koreana resulted in the isolation and identification of a new sesquilignan, named berbikonol (1), along with fourteen known lignan derivatives (2-15) and a new phenolic compound, named berfussinol (16), together with five known ones (17-21). The structures of these new compounds were elucidated on the basis of 1D and 2D NMR spectroscopic data analysis as well as circular dichroism (CD) spectroscopy studies. Compounds 1-5, 7-8, 11, and 14 showed significant cytotoxicity against the XF498 cell line with IC(50) values of 7.14-19.32 μM. In addition, compounds 3-8 and 15 strongly reduced nitric oxide (NO) production in lipopolysaccharide (LPS)-activated BV-2 cells, a microglial cell line.
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
4-Methoxyphenylacetic acid, ReagentPlus®, 99%