MilliporeSigma
  • Activation of GPR35 protects against cerebral ischemia by recruiting monocyte-derived macrophages.
Activation of GPR35 protects against cerebral ischemia by recruiting monocyte-derived macrophages.
Scientific reports (2020-06-12)
Ozayra Sharmin, Ariful Haque Abir, Abdullah Potol, Mahabub Alam, Jewel Banik, A F M Towheedur Rahman, Nuzhat Tarannum, Rasiqh Wadud, Zaki Farhad Habib, Mahbubur Rahman
ABSTRACT
Pamoic acid is a potent ligand for G protein Coupled Receptor 35 (GPR35) and exhibits antinociceptive property. GPR35 activation leads to increased energy utilization and the expression of anti-inflammatory genes. However, its role in brain disorders, especially in stroke, remains unexplored. Here we show in a mouse model of stroke that GPR35 activation by pamoic acid is neuroprotective. Pharmacological inhibition of GPR35 reveals that pamoic acid reduces infarcts size in a GPR35 dependent manner. The flowcytometric analysis shows the expression of GPR35 on the infiltrating monocytes/macrophages and neutrophils in the ischemic brain. Pamoic acid treatment results in a preferential increment of noninflammatory Ly-6CLo monocytes/macrophages in the ischemic brain along with the reduced neutrophil counts. The neuroprotective effect of GPR35 activation depends on protein kinase B (Akt) and p38 MAPK. Together we conclude that GPR35 activation by pamoic acid reprograms Ly-6CLo monocytes/macrophages to relay a neuroprotective signal into the ischemic brain.
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Sigma-Aldrich
ML194, ≥98% (HPLC)