MilliporeSigma
  • Biomimetic niches reveal the minimal cues to trigger apical lumen formation in single hepatocytes.
Biomimetic niches reveal the minimal cues to trigger apical lumen formation in single hepatocytes.
Nature materials (2020-04-29)
Yue Zhang, Richard De Mets, Cornelia Monzel, Vidhyalakshmi Acharya, Pearlyn Toh, Jasmine Fei Li Chin, Noémi Van Hul, Inn Chuan Ng, Hanry Yu, Soon Seng Ng, S Tamir Rashid, Virgile Viasnoff
ABSTRACT
The symmetry breaking of protein distribution and cytoskeleton organization is an essential aspect for the development of apicobasal polarity. In embryonic cells this process is largely cell autonomous, while differentiated epithelial cells collectively polarize during epithelium formation. Here, we demonstrate that the de novo polarization of mature hepatocytes does not require the synchronized development of apical poles on neighbouring cells. De novo polarization at the single-cell level by mere contact with the extracellular matrix and immobilized cadherin defining a polarizing axis. The creation of these single-cell liver hemi-canaliculi allows unprecedented imaging resolution and control and over the lumenogenesis process. We show that the density and localization of cadherins along the initial cell-cell contact act as key triggers of the reorganization from lateral to apical actin cortex. The minimal cues necessary to trigger the polarization of hepatocytes enable them to develop asymmetric lumens with ectopic epithelial cells originating from the kidney, breast or colon.
Sigma-Aldrich
Phalloidin–Atto 565, suitable for fluorescence, ≥80.0% (HPLC)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Ursodeoxycholic acid, ≥99%
Sigma-Aldrich
Trichloro(1H,1H,2H,2H-perfluorooctyl)silane, 97%
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
DAPI, dilactate, ≥98% (HPLC)
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture