MilliporeSigma
  • Synergistic Antifungal Activity of Chitosan with Fluconazole against Candida albicans, Candida tropicalis, and Fluconazole-Resistant Strains.
Synergistic Antifungal Activity of Chitosan with Fluconazole against Candida albicans, Candida tropicalis, and Fluconazole-Resistant Strains.
Molecules (Basel, Switzerland) (2020-11-07)
Wei-Hsuan Lo, Fu-Sheng Deng, Chih-Jung Chang, Ching-Hsuan Lin
ABSTRACT
(1) Background: Few antifungal drugs are currently available, and drug-resistant strains have rapidly emerged. Thus, the aim of this study is to evaluate the effectiveness of the antifungal activity from a combinational treatment of chitosan with a clinical antifungal drug on Candida albicans and Candida tropicalis. (2) Methods: Minimum inhibitory concentration (MIC) tests, checkerboard assays, and disc assays were employed to determine the inhibitory effect of chitosan with or without other antifungal drugs on C. albicans and C. tropicalis. (3) Results: Treatment with chitosan in combination with fluconazole showed a great synergistic fungicidal effect against C. albicans and C. tropicalis, but an indifferent effect on antifungal activity when challenged with chitosan-amphotericin B or chitosan-caspofungin simultaneously. Furthermore, the combination of chitosan and fluconazole was effective against drug-resistant strains. (4) Conclusions: These findings provide strong evidence that chitosan in combination with fluconazole is a promising therapy against two Candida species and its drug-resistant strains.
Sigma-Aldrich
Chitosan, high molecular weight
Pricing and availability is not currently available.
Sigma-Aldrich
Chitosan, low molecular weight
Pricing and availability is not currently available.
Sigma-Aldrich
Chitosan, medium molecular weight
Pricing and availability is not currently available.