MilliporeSigma
  • The human origin recognition complex is essential for pre-RC assembly, mitosis, and maintenance of nuclear structure.
The human origin recognition complex is essential for pre-RC assembly, mitosis, and maintenance of nuclear structure.
eLife (2021-02-02)
Hsiang-Chen Chou, Kuhulika Bhalla, Osama El Demerdesh, Olaf Klingbeil, Kaarina Hanington, Sergey Aganezov, Peter Andrews, Habeeb Alsudani, Kenneth Chang, Christopher R Vakoc, Michael C Schatz, W Richard McCombie, Bruce Stillman
ABSTRACT
The origin recognition complex (ORC) cooperates with CDC6, MCM2-7, and CDT1 to form pre-RC complexes at origins of DNA replication. Here, using tiling-sgRNA CRISPR screens, we report that each subunit of ORC and CDC6 is essential in human cells. Using an auxin-inducible degradation system, we created stable cell lines capable of ablating ORC2 rapidly, revealing multiple cell division cycle phenotypes. The primary defects in the absence of ORC2 were cells encountering difficulty in initiating DNA replication or progressing through the cell division cycle due to reduced MCM2-7 loading onto chromatin in G1 phase. The nuclei of ORC2-deficient cells were also large, with decompacted heterochromatin. Some ORC2-deficient cells that completed DNA replication entered into, but never exited mitosis. ORC1 knockout cells also demonstrated extremely slow cell proliferation and abnormal cell and nuclear morphology. Thus, ORC proteins and CDC6 are indispensable for normal cellular proliferation and contribute to nuclear organization.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Anti-CENP-C Antibody, serum, from rabbit
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Anti-Heterochromatin Protein-1 α Antibody, clone 2HP-1H5, ascites fluid, clone 2HP-1H5, Chemicon®