MilliporeSigma
  • Sulfite Alters the Mitochondrial Network in Molybdenum Cofactor Deficiency.
Sulfite Alters the Mitochondrial Network in Molybdenum Cofactor Deficiency.
Frontiers in genetics (2021-01-26)
Anna-Theresa Mellis, Juliane Roeper, Albert L Misko, Joshua Kohl, Guenter Schwarz
ABSTRACT
Molybdenum cofactor deficiency (MoCD) is an autosomal recessive disorder belonging to the large family of inborn errors in metabolism. Patients typically present with encephalopathy and seizures early after birth and develop severe neurodegeneration within the first few weeks of life. The main pathomechanism underlying MoCD is the loss of function of sulfite oxidase (SO), a molybdenum cofactor (Moco) dependent enzyme located in mitochondrial intermembrane space. SO catalyzes the oxidation of sulfite (SO3 2-) to sulfate (SO4 2-) in the terminal reaction of cysteine catabolism, and in the absence of its activity, sulfurous compounds such as SO3 2-, S-sulfocysteine, and thiosulfate accumulate in patients. Despite growing evidence that these compounds affect neuronal and mitochondrial function, the molecular basis of neuronal dysfunction and cell death in MoCD is still poorly understood. Here we show that mitochondria are severely affected by the loss of SO activity. SO-deficient mouse embryonic fibroblasts display reduced growth rates and impaired ATP production when cultured in galactose, which is an indicator of mitochondrial dysfunction. We also found that mitochondria in SO-deficient cells form a highly interconnected network compared to controls while displaying a slight decrease in motility and unchanged mitochondrial mass. Moreover, we show that the mitochondrial network is directly influenced by SO3 2-, as a moderate elevation of SO3 2- lead to the formation of an interconnected mitochondrial network, while high SO3 2- levels induced fragmentation. Finally, we found a highly interconnected mitochondrial network in MoCD patient-derived fibroblasts, similar to our findings in mouse-derived fibroblasts. We therefore conclude that altered mitochondrial dynamics are an important contributor to the disease phenotype and suggest that MoCD should be included among the mitochondrial disorders.
Sigma-Aldrich
Monoclonal Anti-SUOX antibody produced in mouse, clone 1H1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%