MilliporeSigma
  • Ubiquitin Ligase COP1 Suppresses Neuroinflammation by Degrading c/EBPβ in Microglia.
Ubiquitin Ligase COP1 Suppresses Neuroinflammation by Degrading c/EBPβ in Microglia.
Cell (2020-08-17)
Ada Ndoja, Rohit Reja, Seung-Hye Lee, Joshua D Webster, Hai Ngu, Christopher M Rose, Donald S Kirkpatrick, Zora Modrusan, Ying-Jiun Jasmine Chen, Debra L Dugger, Vineela Gandham, Luke Xie, Kim Newton, Vishva M Dixit
ABSTRACT
Dysregulated microglia are intimately involved in neurodegeneration, including Alzheimer's disease (AD) pathogenesis, but the mechanisms controlling pathogenic microglial gene expression remain poorly understood. The transcription factor CCAAT/enhancer binding protein beta (c/EBPβ) regulates pro-inflammatory genes in microglia and is upregulated in AD. We show expression of c/EBPβ in microglia is regulated post-translationally by the ubiquitin ligase COP1 (also called RFWD2). In the absence of COP1, c/EBPβ accumulates rapidly and drives a potent pro-inflammatory and neurodegeneration-related gene program, evidenced by increased neurotoxicity in microglia-neuronal co-cultures. Antibody blocking studies reveal that neurotoxicity is almost entirely attributable to complement. Remarkably, loss of a single allele of Cebpb prevented the pro-inflammatory phenotype. COP1-deficient microglia markedly accelerated tau-mediated neurodegeneration in a mouse model where activated microglia play a deleterious role. Thus, COP1 is an important suppressor of pathogenic c/EBPβ-dependent gene expression programs in microglia.
Sigma-Aldrich
Calcein Blue, fluorescent dye
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
CDH13 human, recombinant, expressed in E. coli, 0.5 mg protein/mL
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%