MilliporeSigma
  • Macrophage migration inhibitory factor activates the inflammatory response in joint capsule fibroblasts following post-traumatic joint contracture.
Macrophage migration inhibitory factor activates the inflammatory response in joint capsule fibroblasts following post-traumatic joint contracture.
Aging (2021-02-19)
Yuxin Zhang, Shenji Lu, Shuai Fan, Lili Xu, Xin Jiang, Kexin Wang, Bin Cai
ABSTRACT
Joint capsule fibrosis caused by excessive inflammation leading to post-traumatic joint contracture (PTJC). Fibroblasts trigger inflammation under the challenge of various proinflammatory cytokines. Macrophage migration inhibitory factor (MIF) is a prominent proinflammatory cytokine involved in inflammation- and fibrosis-associated pathophysiology, we investigated the role of MIF in PTJC. Using rat PTJC model and fibroblast inflammation model, we detected MIF expression in posterior joint capsule. Primary joint capsule fibroblasts (JFs) were used to investigate the effects of MIF on cell proliferation, migration and proinflammatory cytokines production. The mechanism of JF-mediated events was evaluated by qRT-PCR, western blot and immunoprecipitation. We screened the mRNA expression profile to identify gene candidates that mediate the effect of MIF on JFs. MIF increased in posterior joint capsule following PTJC and co-localized with fibroblasts. Injection of MIF inhibitor significantly suppressed joint capsule inflammation and fibrosis. In vitro, MIF promoted JF proliferation, migration, and inflammation by regulating mitogen-activated protein kinase/nuclear factor-κB pathway through coupling with CD74. Transcriptome analysis revealed that lipid metabolism-related factors Pla2g2a, Angptl4, and Sgpp2, downstream of MIF/CD74, were potentially implicated in JF inflammation. MIF/CD74 axis elicited JF inflammation and may provide new therapeutic targets for joint capsule fibrosis in PTJC.
Sigma-Aldrich
4-IPP, ≥97% (HPLC)