MilliporeSigma
  • High compression deep learning based single-pixel hyperspectral macroscopic fluorescence lifetime imaging in vivo.
High compression deep learning based single-pixel hyperspectral macroscopic fluorescence lifetime imaging in vivo.
Biomedical optics express (2020-11-06)
M Ochoa, A Rudkouskaya, R Yao, P Yan, M Barroso, X Intes
ABSTRACT
Single pixel imaging frameworks facilitate the acquisition of high-dimensional optical data in biological applications with photon starved conditions. However, they are still limited to slow acquisition times and low pixel resolution. Herein, we propose a convolutional neural network for fluorescence lifetime imaging with compressed sensing at high compression (NetFLICS-CR), which enables in vivo applications at enhanced resolution, acquisition and processing speeds, without the need for experimental training datasets. NetFLICS-CR produces intensity and lifetime reconstructions at 128 × 128 pixel resolution over 16 spectral channels while using only up to 1% of the required measurements, therefore reducing acquisition times from ∼2.5 hours at 50% compression to ∼3 minutes at 99% compression. Its potential is demonstrated in silico, in vitro and for mice in vivo through the monitoring of receptor-ligand interactions in liver and bladder and further imaging of intracellular delivery of the clinical drug Trastuzumab to HER2-positive breast tumor xenografts. The data acquisition time and resolution improvement through NetFLICS-CR, facilitate the translation of single pixel macroscopic flurorescence lifetime imaging (SP-MFLI) for in vivo monitoring of lifetime properties and drug uptake.
Sigma-Aldrich
1,1′,3,3,3′,3′-Hexamethylindotricarbocyanine iodide, 97%