MilliporeSigma
  • Synthesis, Antimicrobial and Antioxidant Activities of 2-Isoxazoline Derivatives.
Synthesis, Antimicrobial and Antioxidant Activities of 2-Isoxazoline Derivatives.
Molecules (Basel, Switzerland) (2020-09-24)
Asma Alshamari, Mahmoud Al-Qudah, Fedaa Hamadeh, Lo'ay Al-Momani, Sultan Abu-Orabi
ABSTRACT
A series of derivatives of trans-3-(2,4,6-trimethoxyphenyl)4,5-dihydroisoxazolo-4,5-bis[carbonyl-(4'phenyl)thiosemicarbazide (9) and of trans-3-(2,4,6-trimethoxyphenyl)-4,5-dihydro isoxazolo-4,5-bis(aroylcarbohydrazide) (10a-c) were synthesized from trans-3-(2,4,6-trimethoxyphenyl)-4,5-dihydro-4,5-bis(hydrazenocarbonyl)isoxazole (8). The structures of the compounds were elucidated by both elemental and spectral (IR, NMR, and MS) analysis. Compound 9 shows activity against some bacterial species. No antibacterial activities were observed for compounds 10a-c. The antioxidant activity of the new compounds has been screened. Compound 9 showed higher antioxidant activity using the DPPH (1,1-diphenyl-2-picrylhydrazyl) and ABTS (2'-azino-bis(3-ethylbenzoline-6-sulfonic acid) diammonium salt methods.
Sigma-Aldrich
2,4,6-Trimethoxybenzaldehyde, 98%