MilliporeSigma
Up to 100-fold speed-up and multiplexing in optimized DNA-PAINT.
Nature methods (2020-07-01)
Sebastian Strauss, Ralf Jungmann
ABSTRACT
DNA-PAINT's imaging speed has recently been significantly enhanced by optimized sequence design and buffer conditions. However, this implementation has not reached an ultimate speed limit and is only applicable to imaging of single targets. To further improve acquisition speed, we introduce concatenated, periodic DNA sequence motifs, yielding up to 100-fold-faster sampling in comparison to traditional DNA-PAINT. We extend this approach to six orthogonal sequence motifs, now enabling speed-optimized multiplexed imaging.
Sigma-Aldrich
Dibenzocyclooctyne-maleimide, for Copper-free Click Chemistry
Sigma-Aldrich
SKOV3 Cells GFP-HER2 RFP-EGFR
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Protocatechuate 3,4-Dioxygenase from Pseudomonas sp., lyophilized powder, ≥3 units/mg solid
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)