MilliporeSigma
  • A teaching protocol demonstrating the use of EasyClone and CRISPR/Cas9 for metabolic engineering of Saccharomyces cerevisiae and Yarrowia lipolytica.
A teaching protocol demonstrating the use of EasyClone and CRISPR/Cas9 for metabolic engineering of Saccharomyces cerevisiae and Yarrowia lipolytica.
FEMS yeast research (2019-09-27)
N Milne, L R R Tramontin, I Borodina
ABSTRACT
We present a teaching protocol suitable for demonstrating the use of EasyClone and CRISPR/Cas9 for metabolic engineering of industrially relevant yeasts Saccharomyces cerevisiae and Yarrowia lipolytica, using β-carotene production as a case study. The protocol details all steps required to generate DNA parts, transform and genotype yeast, and perform a phenotypic screen to determine β-carotene production. The protocol is intended to be used as an instruction manual for a two-week practical course aimed at M.Sc. and Ph.D. students. The protocol details all necessary steps for students to engineer yeast to produce β-carotene and serves as a practical introduction to the principles of metabolic engineering including the concepts of boosting native precursor supply and alleviating rate-limiting steps. It also highlights key differences in the metabolism and heterologous production capacity of two industrially relevant yeast species. The protocol is divided into daily experiments covering a two-week period and provides detailed instructions for every step meaning this protocol can be used 'as is' for a teaching course or as a case study for how yeast can be engineered to produce value-added molecules.
Sigma-Aldrich
β-Carotene, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
G 418 disulfate salt solution, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Yeast Nitrogen Base Without Amino Acids, Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization