MilliporeSigma
  • Formulation of PLGA nanoparticles containing short cationic peptide nucleic acids.
Formulation of PLGA nanoparticles containing short cationic peptide nucleic acids.
MethodsX (2020-11-05)
Shipra Malik, Frank J Slack, Raman Bahal
ABSTRACT
Peptide nucleic acids (PNAs) have emerged as one of the most versatile tools with a wide range of biomedical applications including antisense, antimiR, antigene, as well as site-specific gene editing. The application and potential of PNAs has been limited due to low solubility and poor cellular uptake. Several strategies have been employed to overcome the aforementioned challenges like conjugation to cationic peptides or nanotechnology to achieve superior transfection efficiency ex vivo and in vivo. Here, we report a detailed procedure optimized in our lab for synthesis of short cationic PNA probes, which exhibit high purity and yield in comparison to full-length PNA oligomers. We also provide step-by-step details of encapsulating short cationic PNA probes in poly (lactic-co-glycolic acid) nanoparticles by double emulsion solvent evaporation technique. 1.Detailed procedure for synthesis of short cationic PNAs with or without fluorophore (dye) conjugation while ensuring high yield and purity.2.Step-by-step details for encapsulation of short cationic PNAs in PLGA nanoparticles via double emulsion solvent evaporation technique.
Sigma-Aldrich
Boc-Arg(Tos)-OH
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000