MilliporeSigma
Fast antibody fragment motion: flexible linkers act as entropic spring.
Scientific reports (2016-03-30)
Laura R Stingaciu, Oxana Ivanova, Michael Ohl, Ralf Biehl, Dieter Richter
ABSTRACT
A flexible linker region between three fragments allows antibodies to adjust their binding sites to an antigen or receptor. Using Neutron Spin Echo Spectroscopy we observed fragment motion on a timescale of 7 ns with motional amplitudes of about 1 nm relative to each other. The mechanistic complexity of the linker region can be described by a spring model with Brownian motion of the fragments in a harmonic potential. Displacements, timescale, friction and force constant of the underlying dynamics are accessed. The force constant exhibits a similar strength to an entropic spring, with friction of the fragment matching the unbound state. The observed fast motions are fluctuations in pre-existing equilibrium configurations. The Brownian motion of domains in a harmonic potential is the appropriate model to examine functional hinge motions dependent on the structural topology and highlights the role of internal forces and friction to function.
Sigma-Aldrich
Immunoglobulin G from human serum, ≥95% (GE)