MilliporeSigma
  • Mesenchymal Stem Cell/Red Blood Cell-Inspired Nanoparticle Therapy in Mice with Carbon Tetrachloride-Induced Acute Liver Failure.
Mesenchymal Stem Cell/Red Blood Cell-Inspired Nanoparticle Therapy in Mice with Carbon Tetrachloride-Induced Acute Liver Failure.
ACS nano (2018-06-27)
Hongxia Liang, Ke Huang, Teng Su, Zhenhua Li, Shiqi Hu, Phuong-Uyen Dinh, Emily A Wrona, Chen Shao, Li Qiao, Adam C Vandergriff, M Taylor Hensley, Jhon Cores, Tyler Allen, Hongyu Zhang, Qinglei Zeng, Jiyuan Xing, Donald O Freytes, Deliang Shen, Zujiang Yu, Ke Cheng
ABSTRACT
Acute liver failure is a critical condition characterized by global hepatocyte death and often time needs a liver transplantation. Such treatment is largely limited by donor organ shortage. Stem cell therapy offers a promising option to patients with acute liver failure. Yet, therapeutic efficacy and feasibility are hindered by delivery route and storage instability of live cell products. We fabricated a nanoparticle that carries the beneficial regenerative factors from mesenchymal stem cells and further coated it with the membranes of red blood cells to increase blood stability. Unlike uncoated nanoparticles, these particles promote liver cell proliferation in vitro and have lower internalization by macrophage cells. After intravenous delivery, these artificial stem cell analogs are able to remain in the liver and mitigate carbon tetrachloride-induced liver failure in a mouse model, as gauged by histology and liver function test. Our technology provides an innovative and off-the-shelf strategy to treat liver failure.
Sigma-Aldrich
Cell Counting Kit - 8, for quantitation of viable cell number in proliferation and cytotoxicity assays
Sigma-Aldrich
Mouse IL-1 β ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
HSC-T6 Rat Hepatic Stellate Cell Line, The HSC-T6 rat hepatic stellate cell line is a valuable cell model for studies of retinoid metabolism based on their similar retinoid phenotype as primary cells.
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant