MilliporeSigma
  • 2-HG Inhibits Necroptosis by Stimulating DNMT1-Dependent Hypermethylation of the RIP3 Promoter.
2-HG Inhibits Necroptosis by Stimulating DNMT1-Dependent Hypermethylation of the RIP3 Promoter.
Cell reports (2017-06-01)
Zhentao Yang, Bin Jiang, Yan Wang, Hengxiao Ni, Jia Zhang, Jinmei Xia, Minggang Shi, Li-Man Hung, Jingsong Ruan, Tak Wah Mak, Qinxi Li, Jiahuai Han
ABSTRACT
2-hydroxyglutarate-(2-HG)-mediated inhibition of TET2 activity influences DNA hypermethylation in cells harboring mutations of isocitrate dehydrogenases 1 and 2 (IDH1/2). Here, we show that 2-HG also regulates DNA methylation mediated by DNA methyltransferase 1 (DNMT1). DNMT1-dependent hypermethylation of the RIP3 promoter occurred in both IDH1 R132Q knockin mutant mouse embryonic fibroblast (MEFs) and 2-HG-treated wild-type (WT) MEFs. We found that 2-HG bound to DNMT1 and stimulated its association with the RIP3 promoter, inducing hypermethylation that reduces RIP3 protein and consequently impaired RIP3-dependent necroptosis. In human glioma samples, RIP3 protein levels correlated negatively with IDH1 R132H levels. Furthermore, ectopic expression of RIP3 in transformed IDH1-mutated MEFs inhibited the growth of tumors derived from these cells following transplantation into nude mice. Thus, our research sheds light on a mechanism of 2-HG-induced DNA hypermethylation and suggests that impaired necroptosis contributes to the tumorigenesis driven by IDH1/2 mutations.
Sigma-Aldrich
CpGenome Turbo Bisulfite Modification Kit, The CpGenome Turbo Bisulfite Modification Kit is designed to simplify & streamline the bisulfite modification process. In just 90 minutes go from DNA sample to bisulfite converted DNA ready for analysis.