MilliporeSigma
  • COPB2 gene silencing inhibits colorectal cancer cell proliferation and induces apoptosis via the JNK/c-Jun signaling pathway.
COPB2 gene silencing inhibits colorectal cancer cell proliferation and induces apoptosis via the JNK/c-Jun signaling pathway.
PloS one (2020-11-20)
Yan Wang, Guangmei Xie, Min Li, Juan Du, Min Wang
ABSTRACT
Colorectal cancer (CRC) is one of the most common malignant human tumors. It is associated with high morbidity and mortality rates. In recent years, tumor gene therapy has emerged as a promising new approach for colorectal cancer therapy. Herein, we identify and analyze the role of COPB2 (coatomer protein complex, subunit beta 2) in proliferation and apoptosis of CRC cells. To investigate the role of COPB2 in the proliferation and apoptosis of CRC cells, a shCOPB2 vector and a shCtrl vector were constructed for transfection into RKO and HCT116 cells. Cells proliferation was subsequently measured via cell counting kit-8 (CCK8) assay and Celigo cell counting assay. Apoptosis was measured via flow cytometry. The activity level of Caspase 3/7 was measured. Finally, the level of several JNK/c-Jun apoptosis pathway-related proteins were measured to characterize the mechanism of apoptosis. Our results showed that the proliferation rate was decreased and the apoptosis rate was increased in shCOPB2-treated RKO and HCT116 cells compared to those in controls. After the silencing of COPB2, JNK/c-Jun signal pathway activation was increased, the expression levels of apoptosis pathway-related proteins, such as Bad, p53 and Caspase 3, were also increased. COPB2 gene silencing can inhibit RKO and HCT116 cells proliferation and induce apoptosis via the JNK/c-Jun signaling pathway.