MilliporeSigma
  • Neurensin-2 promotes proliferation, invasion and migration of colorectal cancer cells via interaction with SOX12.
Neurensin-2 promotes proliferation, invasion and migration of colorectal cancer cells via interaction with SOX12.
Oncology letters (2020-11-17)
Gang Wang, Kai Yang
ABSTRACT
Colorectal cancer (CRC) is the third most common malignant type of tumor worldwide. Neurensin-2 (NRSN2) is a small neuronal membrane protein associated with tumorigenesis. Therefore, the present study aimed to investigate the association between NRSN2 and CRC, and further examined the underlying mechanism of its effect on CRC metastasis. Human CRC SW620 cells were used to determine the biological functions of NRSN2 in CRC. Cell counting Kit-8 (CCK8), colony formation, wound-healing and transwell assays were performed to evaluate the role of NRSN2 on survival and metastasis of SW620 cells. The interaction between NRSN2 and SOX12 was determined via bioinformatics analysis and confirmed using immunoprecipitation. It was identified that NRSN2 was highly expressed in CRC cells and served a critical role in CRC cell survival compared with in healthy colon epithelial cells. Furthermore, NRSN2-knockdown inhibited the proliferation, invasion and migration of SW620 cells, while NRSN2 overexpression promoted these cellular processes. Additionally, it was demonstrated that NRSN2 could recruit SOX12 in SW620 cells. NRSN2-knockdown decreased SOX12 expression, while NRSN2 overexpression upregulated SOX12 expression. Overall, the present results suggested NRSN2 as a novel biomarker for CRC diagnosis and identified NRSN2 as a potential therapeutic target for CRC treatment.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-SOX12 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
IHC Select Secondary Goat Anti-Rabbit IgG Antibody, prediluted, biotinylated, from goat, biotin conjugate