MilliporeSigma
  • Polyunsaturated fatty acid biosynthesis pathway determines ferroptosis sensitivity in gastric cancer.
Polyunsaturated fatty acid biosynthesis pathway determines ferroptosis sensitivity in gastric cancer.
Proceedings of the National Academy of Sciences of the United States of America (2020-12-09)
Ji-Yoon Lee, Miso Nam, Hye Young Son, Kwangbeom Hyun, Seo Young Jang, Jong Woo Kim, Min Wook Kim, Youngae Jung, Eunji Jang, Seon-Jin Yoon, Jungeun Kim, Jihye Kim, Jinho Seo, Jeong-Ki Min, Kyoung-Jin Oh, Baek-Soo Han, Won Kon Kim, Kwang-Hee Bae, Jaewhan Song, Jaehoon Kim, Yong-Min Huh, Geum-Sook Hwang, Eun-Woo Lee, Sang Chul Lee
ABSTRACT
Ferroptosis is an iron-dependent regulated necrosis mediated by lipid peroxidation. Cancer cells survive under metabolic stress conditions by altering lipid metabolism, which may alter their sensitivity to ferroptosis. However, the association between lipid metabolism and ferroptosis is not completely understood. In this study, we found that the expression of elongation of very long-chain fatty acid protein 5 (ELOVL5) and fatty acid desaturase 1 (FADS1) is up-regulated in mesenchymal-type gastric cancer cells (GCs), leading to ferroptosis sensitization. In contrast, these enzymes are silenced by DNA methylation in intestinal-type GCs, rendering cells resistant to ferroptosis. Lipid profiling and isotope tracing analyses revealed that intestinal-type GCs are unable to generate arachidonic acid (AA) and adrenic acid (AdA) from linoleic acid. AA supplementation of intestinal-type GCs restores their sensitivity to ferroptosis. Based on these data, the polyunsaturated fatty acid (PUFA) biosynthesis pathway plays an essential role in ferroptosis; thus, this pathway potentially represents a marker for predicting the efficacy of ferroptosis-mediated cancer therapy.
Sigma-Aldrich
ML 210, ≥98% (HPLC)
Sigma-Aldrich
CP-24879 hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-FADS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
SC-26196, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Liproxstatin-1, >98% (HPLC)