MilliporeSigma
  • Evaluation of bisulfite kits for DNA methylation profiling in terms of DNA fragmentation and DNA recovery using digital PCR.
Evaluation of bisulfite kits for DNA methylation profiling in terms of DNA fragmentation and DNA recovery using digital PCR.
PloS one (2018-06-15)
Sam Kint, Ward De Spiegelaere, Jonas De Kesel, Linos Vandekerckhove, Wim Van Criekinge
ABSTRACT
DNA methylation is one of the most important epigenetic modifications in the regulation of gene transcription. The current gold standard to study this modification is bisulfite sequencing. Although multiple commercial bisulfite treatment kits provide good conversion efficiencies, DNA loss and especially DNA fragmentation remain troublesome. This hampers DNA methylation profiling of long DNA sequences. Here, we explored the performance of twelve commercial bisulfite kits by an in-depth comparison of DNA fragmentation using gel electrophoresis, qPCR and digital PCR, DNA recovery by spectroscopic measurements and digital PCR and conversion efficiency by next generation sequencing. The results show a clear performance difference between the bisulfite kits, and depending on the specific goal of the study, the most appropriate kit might differ. Moreover, we demonstrated that digital PCR is a valuable method to monitor both DNA fragmentation as well as DNA recovery after bisulfite treatment.
Sigma-Aldrich
Imprint® DNA Modification Kit, For bisulfite DNA conversion & purification
Sigma-Aldrich
CpGenome Turbo Bisulfite Modification Kit, The CpGenome Turbo Bisulfite Modification Kit is designed to simplify & streamline the bisulfite modification process. In just 90 minutes go from DNA sample to bisulfite converted DNA ready for analysis.