MilliporeSigma
  • Antioxidant and Antiapoptotic Polyphenols from Green Tea Extract Ameliorate CCl4-Induced Acute Liver Injury in Mice.
Antioxidant and Antiapoptotic Polyphenols from Green Tea Extract Ameliorate CCl4-Induced Acute Liver Injury in Mice.
Chinese journal of integrative medicine (2019-11-27)
Jian-Xin Diao, Jin-Ying Ou, Huan Dai, Hai-Ye Li, Wei Huang, He-Yu Hua, Ting Xie, Ming Wang, Yun-Gao Yang
ABSTRACT
To investigate the phenolic composition, antioxidant properties, and hepatoprotective mechanisms of polyphenols from green tea extract (GTP) in carbon tetrachloride (CCl4)-induced acute liver injury mouse model. High-performance liquid chromatography was used to analyze the chemical composition of the extract. Antioxidant activity of GTP was assessed by O2∙-, OH∙, DPPH∙, and ferric-reducing antioxidant power (FRAP) assay in vitro. Sixty Kunming mice were divided into 6 groups including control, model, low-, medium-, and high-doses GTP (200, 400, 800 mg/kg) and vitamin E (250 mg/kg) groups, 10 in each group. GTP and vitamin E were administered at a level of abovementioned doses twice per day for 7 days prior to exposure to a single injection of CCl4. Hepatoprotective effects of GTP were evaluated in a CCl4-induced mouse model of acute liver injury, using commercial enzyme linked immunosorbent assay kits, histopathological observation, terminal deoxynucleotidyl transferase-mediated dUTPNick-end labeling (TUNEL) assay and Western blot. GTP contained 98.56 µg gallic acid equivalents per milligram extract total polyphenols, including epicatechingallate, epigallocatechin gallate, epicatechin, and epigallocatechin. Compared with the model group, low-, medium-, or high doses GTP significantly decreased serum levels of alanine aminotransferase and aspartate transaminase (P<0.01). Histopathological observation confirmed that pretreatment of GTP prevented swelling and necrosis in CCl4-exposed hepatocytes. Hepatoprotective effects of low-, medium-, and high-dose GTP were associated with eliminating free radicals and improving superoxide dismutase, catalase, and glutathione peroxidase activity in the liver. Additionally, low-, medium-, and high-dose GTP decreased cell apoptosis in the CCl4-exposed liver (P<0.01). Phosphorylated nuclear factor kappa-B (NF-κB), p53, Bcl-2 associated x protein/B-cell lymphoma/leukemia-2 gene, cytochrome C, and cleaved caspase-3 levels were downregulated compared with the model group (P<0.01). GTP achieves hepatoprotective effects by improving hepatic antioxidant status and preventing cell apoptosis through caspase-3-dependent signaling pathways.
Sigma-Aldrich
p-Nitroblue Tetrazolium Chloride, NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC₅₀ = 3-4 µM).
Sigma-Aldrich
Phenazine methosulfate
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
2,2-Di(4-tert-octylphenyl)-1-picrylhydrazyl, free radical