MilliporeSigma
  • S100A9 promotes prostate cancer cell invasion by activating TLR4/NF-κB/integrin β1/FAK signaling.
S100A9 promotes prostate cancer cell invasion by activating TLR4/NF-κB/integrin β1/FAK signaling.
OncoTargets and therapy (2020-09-05)
Zhonghua Lv, Wenlin Li, Xichao Wei
ABSTRACT
S100A9, which is expressed in prostate cancer, has been reported in association with prostate cancer progression. However, the role of S100A9 in prostate cancer metastasis is largely unknown. The aim of this study was to investigate the effect of S100A9 on prostate cancer cell invasion and the involved mechanisms. Integrin β1 expression in PC-3 and DU-145 cells was determined by quantitative real-time polymerase chain reaction (PCR) (qRT-PCR) and Western blot. Cellular invasion was measured by transwell invasion assay. Western blot was used to determine protein expression. Concentrations of S100A9 and fibronectin were analyzed by enzyme-linked immunosorbent assay. The protein interaction was detected by immunoprecipitation. The NF-κB activity was measured by luciferase reporter assay. The DU-145 cells metastasis in vivo was determined in mice xenograft models after S100A9 overexpression. S100A9 promoted prostate cancer cells invasion, integrin β1 expression and fibronectin secretion. Further investigation evidenced that S100A9 interacted with Toll-like receptor 4 (TLR4) and activated NF-κB, which was responsible for tumor cell invasion, integrin β1 up-regulation and focal adhesion kinase (FAK) phosphorylation. Furthermore, integrin β1 inhibition led to decreased FAK phosphorylation and reduced tumor cell invasion. Overexpression of S100A9 increased xenograft tumor micro-metastases, integrin β1 expression and induced NF-κB and FAK activation in vivo. Our study demonstrated that S100A9 promotes prostate cancer cell invasion, and one of the underlying molecular mechanisms is that S100A9 activates integrin β1/FAK through TLR4/NF-κB signaling leading to metastasis of prostate cancer cell.
Sigma-Aldrich
Monoclonal Anti-Fibronectin, Cellular antibody produced in mouse, clone FN-3E2, ascites fluid
Sigma-Aldrich
Human Fibronectin ELISA Kit, for cell culture supernatants, plasma, and serum samples
Sigma-Aldrich
PF-562,271, ≥98% (HPLC)