MilliporeSigma
  • Variation in HIV-1 Nef function within and among viral subtypes reveals genetically separable antagonism of SERINC3 and SERINC5.
Variation in HIV-1 Nef function within and among viral subtypes reveals genetically separable antagonism of SERINC3 and SERINC5.
PLoS pathogens (2020-09-15)
Steven W Jin, Francis M Mwimanzi, Jaclyn K Mann, Mwebesa Bosco Bwana, Guinevere Q Lee, Chanson J Brumme, Peter W Hunt, Jeff N Martin, David R Bangsberg, Thumbi Ndung'u, Zabrina L Brumme, Mark A Brockman
ABSTRACT
HIV Nef counteracts cellular host restriction factors SERINC3 and SERINC5, but our understanding of how naturally occurring global Nef sequence diversity impacts these activities is limited. Here, we quantify SERINC3 and SERINC5 internalization function for 339 Nef clones, representing the major pandemic HIV-1 group M subtypes A, B, C and D. We describe distinct subtype-associated hierarchies for Nef-mediated internalization of SERINC5, for which subtype B clones display the highest activities on average, and of SERINC3, for which subtype B clones display the lowest activities on average. We further identify Nef polymorphisms that modulate its ability to counteract SERINC proteins, including substitutions in the N-terminal domain that selectively impair SERINC3 internalization. Our findings demonstrate that the SERINC antagonism activities of HIV Nef differ markedly among major viral subtypes and between individual isolates within a subtype, suggesting that variation in these functions may contribute to global differences in viral pathogenesis.
Sigma-Aldrich
Duolink® flowPLA Mouse/Rabbit Starter Kit - Far Red, Duolink® PLA kit for Flow Cytometry with Far Red Detection and Mouse/Rabbit probes